Graphite tubes for atomic absorption
Pyrolytically coated partition graphite tube
Katalog # PERCN9304625
Leverantör: PerkinElmer
Gå till:
Produktdetaljer och dokument
Graphite tubes made with highest quality material.
Suitable AA instruments.
Specifikationer
- Beskrivning:Pyrolytically coated partition graphite tube