Microscopy scissors
Blunt, straight, 125 mm
Katalog # RSGA101.125
Leverantör: RSG
Gå till:
Product Details & Documents
Carbon steel, nickel plated, 8/10 mm.
Specifikationer
- Typ:Blunt, straight
- Längd:125 mm
- Seq:02