Brushes, FlexBrush™
S-Brush adapter, S-shaped brush, stainless steel with plastic handle, Nylon, 57,1 mm, Rostfritt stål, 533 mm
Catalog # MICRBRWM-22512S_U
Supplier: MICRONOVA
Jump to:
Product Details & Documents
The FlexBrush™ is ideal for cleaning over and around piping and fittings. The flexibility of the head allows the user to shape the head for custom cleaning applications.
- Bendable brush with polyester or microfibre cover
- Acme threaded end attaches to a variety of handles for extended reach
- Threaded handle (STA series) can be added for extended reach
- Covers are compatible with strong disinfectants and autoclavable
Specifications
- Borstmaterial:Nylon
- Borsthuvud Ø:57,1 mm
- Handle material:Rostfritt stål
- Beskrivning:S-Brush adapter, S-shaped brush, stainless steel with plastic handle
- Seq:02
- Total längd:533 mm