Multipacks, Tacta®
3-pack MAX, incl. three single channel pipettes (100 - 1000 / 500 - 5000 / 1000 - 10000 µl), Optifit tips (10 - 1000 μl (96/tray) / 0,1 - 5 ml, bulk (100) / 1 - 10 ml, bulk (50)) and one linear stand
Katalog # 613-5633
Leverantör: Sartorius
Gå till:
Product Details & Documents
The Tacta® Multipacks offer an opportunity to test and get started with Tacta® mechanical pipettes. Tacta® mechanical pipette offers excellent performance and safety in manual pipetting. It is designed to maintain high accuracy and precision in repetitive and long-lasting pipetting series.
- Reliable results, even over lengthy pipetting periods
- Controlled and smooth tip ejection
- The Optilock® system provides flexibility for volume adjustment and locking
There are different multipack options to choose from ranging from a set of 3 to a set of 5 pipettes.
Beställningsinformation: All sets come with a linear stand for the safe storage of your pipettes as well as suitable racked or bulk-packed Optifit tips.
Specifikationer
- Beskrivning:3-pack MAX, incl. three single channel pipettes (100 - 1000 / 500 - 5000 / 1000 - 10000 µl), Optifit tips (10 - 1000 μl (96/tray) / 0,1 - 5 ml, bulk (100) / 1 - 10 ml, bulk (50)) and one linear stand
- Seq:08
- Cat. No.:613-5633
- Miljöcertificering:Renewable energy certificate; ISO 14001
- Environmentally Preferable:Low Manufacturing Impact