Order Entry
Sweden
Orders LinkContactUsLinkComponent
Fluorbensen ≥99.0%

Fluorbensen ≥99.0%

100 g
Catalog # TCIAF0034-100G
Flammable
Irritant
Health
Supplier:   TCI
Fluorbensen ≥99.0%
Catalog # TCIAF0034-100G
Supplier:  TCI
CAS Number:  462-06-6
Fluorbensen ≥99.0%
Fluorbensen ≥99.0%

100 g

CAS Number:  462-06-6
Price
565,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Phenylfluoride
  • CAS Number:462-06-6
  • Formula:C₆H₅F
  • MDL Number:MFCD00000280
  • Molecular Weight:96.1 g/mol
  • Merck Index:14,04170
  • Boiling Pt:85°C
  • Melting Pt:–41 °C
  • Density:1,023 g/cm³ (20 °C)
  • Flash Pt:–13°C
  • Flash Pt:-13°C
  • Storage Temperature:Omgivande
  • EINECS:207-321-7
  • UN:2387
  • ADR:3,II
Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

Specifications

  • Förp.:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.