Order Entry
Sweden
Orders LinkContactUsLinkComponent
Buffert, bruksfärdig

Buffert, bruksfärdig

Technical buffer TEP-10 Trace/10, 10,01, Plastic bottle, 10
Katalog # WTWA108809
Leverantör:   WTW
Buffert, bruksfärdig
Katalog # WTWA108809
Leverantör:  WTW
CAS-nummer:  
Buffert, bruksfärdig
Buffert, bruksfärdig

Technical buffer TEP-10 Trace/10, 10,01, Plastic bottle, 10

CAS-nummer:  
Price
3 100,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 10 Items
Produktdetaljer och dokument

Every bottle has its own lot reference with the expiration date.

  • Shelf life up to 3 years (unopened)
  • Reference temperature at 25 °C
  • Economic consumption
  • Easiest handling by use of the dosing vessel
  • One litre container with a separate compartment for the sample

Accuracy is ±0,03 pH

Certifications: Traceable to NIST.

Specifikationer

Specifikationer

  • Förpackningstyp:
    Plastic bottle
  • Förp.:
    10
  • Beskrivning:
    Technical buffer TEP-10 Trace/10
  • pH-värde:
    10,01

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.