Orders LinkContactUsLinkComponent
Column packing support, Quickfit®
29 mm, 34/35 NS
Katalog # BIBBEX13/25
Leverantör:   SciLabware
Column packing support, Quickfit®
Katalog # BIBBEX13/25
Leverantör:  SciLabware
Column packing support, Quickfit®
Column packing support, Quickfit®
29 mm, 34/35 NS
Price
1 050,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 5 Items
Product Details & Documents
PYREX®, borosilicate glass.
Designed to support Raschig rings within plain fractionating columns. Support is passed through socket of column and rests at the constriction above the cone.
Specifikationer
  • Kärna:
    34/35 NS
  • Ø:
    29 mm
  • Seq:
    03