Orders LinkContactUsLinkComponent
Test kits, drop count tests
Test kits, drop count tests
Carbon dioxide, reagent set for model CA-23, drop count titration, set includes phenolphthalein indicator solution and sodium hydroxide standard solution (0,01 N)., 1 KIT
Katalog # HACH2437500
Leverantör:   Hach
Test kits, drop count tests
Katalog # HACH2437500
Leverantör:  Hach
CAS-nummer:  
Test kits, drop count tests
Test kits, drop count tests
Carbon dioxide, reagent set for model CA-23, drop count titration, set includes phenolphthalein indicator solution and sodium hydroxide standard solution (0,01 N)., 1 KIT
CAS-nummer:  
Price
605,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 KIT
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit
Varning: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifikationer
Specifikationer
  • Förp.:
    1 KIT
  • Beskrivning:
    Carbon dioxide, reagent set for model CA-23, drop count titration, set includes phenolphthalein indicator solution and sodium hydroxide standard solution (0,01 N).