Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
TALLPREP METAL BEAD LYS. MATRIX T 4.5 ML 1 * 100 Items
ICNA116945100
:
MP Biomedicals
TALLPREP METAL BEAD LYS. MATRIX T 4.5 ML 1 * 100 Items
ICNA116945100
:
MP Biomedicals