Orders LinkContactUsLinkComponent
Sterilisation Boxes for Pipettes
Pipette box for the storage of pipettes (235 - 305 mm)
Katalog # BRND28925
Leverantör:   Brand
Sterilisation Boxes for Pipettes
Katalog # BRND28925
Leverantör:  Brand
Sterilisation Boxes for Pipettes
Sterilisation Boxes for Pipettes
Pipette box for the storage of pipettes (235 - 305 mm)
Price
920,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Produktdetaljer och dokument
These pipette boxes made of aluminum are ideal for the storage and sterilisation of pipettes with different lengths.
  • With inner silicone coating
  • Very good temperature resistance
  • Very good chemical resistance
Specifikationer
  • Beskrivning:
    Pipette box for the storage of pipettes (235 - 305 mm)
  • Seq:
    02