Order Entry
Sweden
Orders LinkContactUsLinkComponent
Accessories for centrifuges

Accessories for centrifuges

Adapter (reducer), boring Ø×L: 17,5×60 mm, for 5×4 - 10 ml tubes (Ø 15/16 mm), for use with carrier 1560 in rotor 1554
Katalog # 521-2222
   Low Manufacturing Impact
Leverantör:   Hettich
Accessories for centrifuges
Katalog # 521-2222
Leverantör:  Hettich
   Low Manufacturing Impact
Accessories for centrifuges
Accessories for centrifuges

Adapter (reducer), boring Ø×L: 17,5×60 mm, for 5×4 - 10 ml tubes (Ø 15/16 mm), for use with carrier 1560 in rotor 1554

Price
860,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivning:
    Adapter (reducer), boring Ø×L: 17,5×60 mm, for 5×4 - 10 ml tubes (Ø 15/16 mm), for use with carrier 1560 in rotor 1554
  • Seq:
    08
  • Cat. No.:
    521-2222
  • Miljöcertificering:
    ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.