Single-use bags for HYPERStack® vessels
Trypsin bag, 5 L
Supplier: Corning
Jump to:
Product Details & Documents
These single-use bags can be connected to tubing via tube welding or by using the pre-assembled multipurpose connectors.
They can be used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution.
Documents
Specifications
- Kapacitet:5 L
- Beskrivning:Trypsin bag
- Seq:01