Orders LinkContactUsLinkComponent
Separating funnel, globe shaped
Separating funnel, globe shaped
1000 ml, 29/32 NS
Catalog # KAVA426351940
Supplier:   Kavalier
Separating funnel, globe shaped
Catalog # KAVA426351940
Supplier:  Kavalier
Separating funnel, globe shaped
Separating funnel, globe shaped
1000 ml, 29/32 NS
Price
3 200,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • Very good temperature resistance
Suitable for separation of particular phases of the mixture. With standard ground joint stopper.
Specifications
  • NS-hylsa:
    29/32 NS
  • Kapacitet:
    1000 ml
  • Seq:
    02