Orders LinkContactUsLinkComponent
Plastic Additive, Reference Standards
Plastic Additive, Reference Standards
Plastic Additive 2, USP reference standard, Vial, 200 mg
Katalog # 1544938.
Leverantör:   USP
Plastic Additive, Reference Standards
Katalog # 1544938.
Leverantör:  USP
CAS-nummer:  
Plastic Additive, Reference Standards
Plastic Additive 2, USP reference standard, Vial, 200 mg
CAS-nummer:  
Price
4 275,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 200 mg
Produktdetaljer och dokument
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Plastic Additive 1 (Ethylene bis[3,3-bis[3-(1,1-dimethylethyl)-4-hydroxyphenyl] butanoate]) CAS 32509-66-3
Plastic Additive 2 (pentaerythrityl tetrakis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]) CAS 6683-19-8
Plastic Additive 3 (2,2',2'',6,6',6''-hexa-tert-butyl-4,4',4''-[(2,4,6-trimethyl-1,3,5-benzenetriyl)trismethylene]triphenol) CAS 1709-70-2
Plastic Additive 4 (Octadecyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate) CAS 2082-79-3
Plastic Additive 5 (Tris(2,4-di-tert-butylphenyl) phosphite) CAS 31570-04-4
Plastic Additive 6 (1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-s-triazine-2,4,6(1H,3H,5H)-trione) CAS 27676-62-6
Plastic Additive 8 (Dioctadecyl disulfide) CAS 2500-88-1
Plastic Additive 9 (Didodecyl 3,3'-thiodipropionate) CAS 123-28-4
Plastic Additive 10 (dioctadecyl 3,3'-thiodipropionate) CAS 693-36-7
Plastic Additive 11 (Copolymer of Dimethyl Succinate and (4-hydroxy-2,2,6,6-tetramethylpiperidin-1-yl)ethanol)) CAS 65447-77-0
Plastic Additive 12 (Oleamide) CAS 301-02-0
Plastic Additive 13 (Erucamide) CAS 112-84-5
Plastic Additive 14 (Di(2-ethylhexyl) phthalate) CAS 117-81-7
Plastic Additive 15 (Epoxidized Soya Oil) CAS 8013-07-8
Plastic Additive 16 (Epoxidized Linseed Oil) CAS 8016-11-3
Certifications: US Pharmacopea Reference Standard
Specifikationer
Specifikationer
  • Förpackningstyp:
    Vial
  • Förp.:
    200 mg
  • Standard:
    USP reference standard
  • Beskrivning:
    Plastic Additive 2
Specifikationer
Rekommendationer
Rekommendationer anpassas endast baserat på dina shoppingpreferenser om du har gett ditt samtycke genom att aktivera "Förbättra min shoppingupplevelse" på sidan "Personlig information".
Annars får du generiska rekommendationer.