Basic Violet 3 ACS
Glass bottle, 50 g
Leverantör: Thermo Fisher Scientific
Gå till:
Product Details & Documents
Synonymer: 4-[4,4'-Bis(dimetylamino) benzhydryliden]cyklohexa-2,5-dien-1-ylidendimetylammoniumklorid, CI 42555
- CAS-nummer:548-62-9
- Formel:C₂₅H₃₀ClN₃
- MDL-nummer:MFCD00011750
- Molecular Weight:407.99 g/mol
- Smält Pt:173 °C (dec.)
- Densitet:1,19 g/cm³ (20 °C)
- Förvaringstemperatur:Omgivande
- FN:3077
- ADR:9,III
Documents
ACRO44757
SDS
Specifikationer
Specifikationstestresultat
- Appearance:Dark green Powder or crystals
- Assay:≥90.0 % (on dry substance)
- Loss on drying:≤7.5 %
- Absorbance characteristics:(0.98-1.9) Passes test
- Sensitivity:Passes test
Specifikationer
- Förpackningstyp:Glass bottle
- Förp.:50 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



