Kiselgel 99.998%
Glass bottle, 100 g
Leverantör: Thermo Fisher Scientific
Gå till:
Produktdetaljer och dokument
Synonymer: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
- CAS-nummer:14808-60-7
- Formel:SiO₂
- MDL-nummer:MFCD00011232
- Molecular Weight:60.08 g/mol
- Kokande Pt:2230 °C (1013 hPa)
- Smält Pt:1610 °C
- Densitet:2,196 g/cm³ (20 °C)
- Förvaringstemperatur:Omgivande
Specifikationer
Specifikationstestresultat
- Appearance:White Crystalline powder or crystals
- Assay:44.5 to 47.9 % (Si)
- Total trace metal impurities:≤20 ppm
Specifikationer
- Förpackningstyp:Glass bottle
- Förp.:100 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



