Kiselgel 99.998%
Glass bottle, 20 g
: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
- :14808-60-7
- :SiO₂
- :MFCD00011232
- :60.08 g/mol
- :2230 °C (1013 hPa)
- :1610 °C
- :2,196 g/cm³ (20 °C)
- :Omgivande
- Appearance:White Crystalline powder or crystals
- Assay:44.5 to 47.9 % (Si)
- Total trace metal impurities:≤20 ppm
- Förpackningstyp:Glass bottle
- Förp.:20 g



