Kiselgel 99.998%
Glass bottle, 5 g
: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
- :14808-60-7
- :SiO₂
- :MFCD00011232
- :60.08 g/mol
- :2230 °C (1013 hPa)
- :1610 °C
- :2,196 g/cm³ (20 °C)
- :Omgivande
Documents
ACRO43715
SDS
- Appearance:White Crystalline powder or crystals
- Assay:44.5 to 47.9 % (Si)
- Total trace metal impurities:≤20 ppm
- Förpackningstyp:Glass bottle
- Förp.:5 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



