Basic Violet 3, ren indicator
Glass bottle, 100 g
Leverantör: Thermo Fisher Scientific
Basic Violet 3, ren indicator
Katalog # ACRO212121000
Leverantör: Thermo Fisher Scientific
CAS-nummer: 548-62-9
Gå till:
Product Details & Documents
Synonymer: 4-[4,4'-Bis(dimetylamino) benzhydryliden]cyklohexa-2,5-dien-1-ylidendimetylammoniumklorid, CI 42555
- CAS-nummer:548-62-9
- Formel:C₂₅H₃₀ClN₃
- MDL-nummer:MFCD00011750
- Molecular Weight:407.99 g/mol
- Merck Index:13,04406
- Smält Pt:173 °C (dec.)
- Densitet:1,19 g/cm³ (20 °C)
- Förvaringstemperatur:Omgivande
- EINECS:208-953-6
- FN:3077
- ADR:9,III
Documents
Specifikationer
Specifikationstestresultat
- Appearance:Dark green Crystalline powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
- Sensitivity:(as indicator) Passes test
Specifikationer
- Förpackningstyp:Glass bottle
- Förp.:100 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



