Accessories
Flexible discharge tube with recirculation valve, PTFE, coiled, length approx 800 mm, with safety handle, for Dispensette® S/S Organic 2,5+10 ml
Katalog # 613-5221
Leverantör:   Brand
Accessories
Katalog # 613-5221
Leverantör:  Brand
Accessories
Accessories
Flexible discharge tube with recirculation valve, PTFE, coiled, length approx 800 mm, with safety handle, for Dispensette® S/S Organic 2,5+10 ml
Price
1 275,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Specifikationer
  • Beskrivning:
    Flexible discharge tube with recirculation valve, PTFE, coiled, length approx 800 mm, with safety handle, for Dispensette® S/S Organic 2,5+10 ml
  • Seq:
    21