Orders LinkContactUsLinkComponent
Osmolality standards
Osmolality standards
Osmolality standard, 1000 mOsm/kg H₂O, Ampoule, 12×5 ml, 1 KIT
Katalog # REGERE-OSM-1000
Leverantör:   REAGECON
Osmolality standards
Katalog # REGERE-OSM-1000
Leverantör:  REAGECON
CAS-nummer:  
Osmolality standards
Osmolality standard, 1000 mOsm/kg H₂O, Ampoule, 12×5 ml, 1 KIT
CAS-nummer:  
Price
2 325,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 KIT
Produktdetaljer och dokument
Osmolality standards provide ease of use when calibrating all types of osmometers. All Osmolality standards are manufactured using high purity raw materials. These products are prepared gravimetrically on a weight/weight basis. Both solute (salts) and solvent (water) are weighed on a balance calibrated by using OIML traceable weights.
  • Ready to use and presented in convenient ampoules
  • Extended shelf life and has low uncertainty of measurement
  • Consistency of product-Independent, traceable
  • Consist of 12 ampoules of 5 ml
The Osmolality of the standard is verified using a high performance calibrated, temperature controlled osmometer. The calibration of this instrument is completed using high purity certified Osmolality standards.
Certifications: ISO17025 accreditation for calibration of non automatic weighing machines (INAB ref: 265C). The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.
Dokument
Specifikationer
Specifikationer
  • Förpackningstyp:
    Ampoule
  • Förp.:
    1 KIT
  • Beskrivning:
    Osmolality standard
  • Pk Info:
    12×5 ml
  • Osmolality:
    1000 mOsm/kg H₂O