Trays
Flask tray with threaded holes to attach 25 to1000 ml SC flask clamps, SH plate holders and SR racks, for OLS26
Katalog # 462-0539
Leverantör: GRANT INSTRUMENTS
Gå till:
Produktdetaljer och dokument
Specifikationer
- Beskrivning:Flask tray with threaded holes to attach 25 to1000 ml SC flask clamps, SH plate holders and SR racks, for OLS26