Orders LinkContactUsLinkComponent
Trays
Trays
Test tube tray, compatible with SR test tube racks or can be used alone to accommodate bags and miscellaneous vessels, for LSB18 (holds up to 5× SR test tube racks)
Katalog # 462-0541
Leverantör:   GRANT INSTRUMENTS
Trays
Katalog # 462-0541
Leverantör:  GRANT INSTRUMENTS
Trays
Trays
Test tube tray, compatible with SR test tube racks or can be used alone to accommodate bags and miscellaneous vessels, for LSB18 (holds up to 5× SR test tube racks)
Price
4 550,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Produktdetaljer och dokument
Specifikationer
  • Beskrivning:
    Test tube tray, compatible with SR test tube racks or can be used alone to accommodate bags and miscellaneous vessels, for LSB18 (holds up to 5× SR test tube racks)
  • Seq:
    05