Trays
Stainless steel perforated base tray, allows bath to be used as an unstirred bath, for LSB18/OLS26
Katalog # 462-0521
Leverantör: GRANT INSTRUMENTS
Gå till:
Product Details & Documents
Specifikationer
- Beskrivning:Stainless steel perforated base tray, allows bath to be used as an unstirred bath, for LSB18/OLS26
- Seq:11