Screw caps
Caps, PP, white, LDPE foam lined, 48-400
Katalog # EPSC067-4800WBP
Leverantör: EP SCIENTIFIC
Gå till:
Produktdetaljer och dokument
LDPE-lined white caps.
- For general-purpose HDPE bulk separate containers
- Same high quality caps used on certified HDPE environmental sample containers
Beställningsinformation: Bottles sold separately (IR013-series and IR012-series).
Specifikationer
- Gänga:48-400
- Beskrivning:Caps, PP, white, LDPE foam lined
- Seq:04
- Cat. No.:EPSC067-4800WBP
- Miljöcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact