[EN]TUBE-MINIMUM UNION 1INPL PrimeLock(T 1 * 1 Items
Katalog # ENTEPSMT-16-3X
Leverantör: ENTEGRIS
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Leverantör: VWR Chemicals
Förseglingsfilm för manuell applicering, Parafilm® M
Leverantör: HEATHROW
Metanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade för HPLC
Leverantör: VWR Chemicals
Vatten, HiPerSolv CHROMANORM® för HPLC
Leverantör: VWR Chemicals
VWR® NITRILE LIGHT, Nitrilhandskar
Leverantör: VWR Collection
VWR®, Transferpipetter, hög prestanda
Leverantör: VWR Collection
Handskar, nitril, Kimtech™ Purple Nitrile™
Leverantör: KIMBERLY CLARK
VWR®, Serologisk pipett, standard
Leverantör: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverantör: VWR Chemicals
VWR®, Nitrilhandskar
Leverantör: VWR Collection
VWR®, Ansiktsservett
Leverantör: VWR Collection
Teststrips, MQuant®, Supelco®
Leverantör: Merck