Diffraction gratings
Diaphragm with 4 multiple slits, Slit spacing: 0,25 mm, Slit width: 0, 15 mm
BINHU14102
Marketsource
Photographic diffraction objects in slide frames.
- Dimensions: 50×50 mm
- Beskrivning:Diaphragm with 4 multiple slits, Slit spacing: 0,25 mm, Slit width: 0, 15 mm