Diffraction gratings
Diaphragm with 4 double slits, Slit spacing: 0,15 mm, Slit width: 0,25 mm; 0,50 mm; 0,75 mm; 1,00 mm
Katalog # BINHU14101
Marketsource
Leverantör: 3B Scientific
Gå till:
Produktdetaljer och dokument
Photographic diffraction objects in slide frames.
- Dimensions: 50×50 mm
Specifikationer
- Beskrivning:Diaphragm with 4 double slits, Slit spacing: 0,15 mm, Slit width: 0,25 mm; 0,50 mm; 0,75 mm; 1,00 mm