Diffraction gratings
Diaphragm with 3 double slits, Slit spacing: 0,30 mm, Slit width: 0,10 mm; 0,15 mm; 0,20 mm
Katalog # BINHU14100
Marketsource
Leverantör: 3B Scientific
Gå till:
Produktdetaljer och dokument
Photographic diffraction objects in slide frames.
- Dimensions: 50×50 mm
Specifikationer
- Beskrivning:Diaphragm with 3 double slits, Slit spacing: 0,30 mm, Slit width: 0,10 mm; 0,15 mm; 0,20 mm