Orders LinkContactUsLinkComponent
VWR® VisiScope®, Accessories
VWR® VisiScope®, Accessories
Phase contrast set, complete, incl. W-Plan objectives (10×, 40×, 100×) and BF/DF/PH condenser
Catalog # 630-2058
Supplier:   VWR Collection
VWR® VisiScope®, Accessories
Catalog # 630-2058
Supplier:  VWR Collection
VWR® VisiScope®, Accessories
VWR® VisiScope®, Accessories
Phase contrast set, complete, incl. W-Plan objectives (10×, 40×, 100×) and BF/DF/PH condenser
Price
12 550,00 kr
Log in for availability
1 * 1 Items
Specifications
  • Beskrivning:
    Phase contrast set, complete, incl. W-Plan objectives (10×, 40×, 100×) and BF/DF/PH condenser
  • Seq:
    82