VWR®, N-Plan Objectives
N-plan objective (oil/water), 100×/1,25 mm (i)
Katalog # 630-2009
Leverantör: VWR Collection
Gå till:
Product Details & Documents
These objectives are ideal for use in the education field and for laboratory routine applications.
IOS N-Plan objectives feature an infinity-corrected optical system.
Documents
VWR® for Microscopy
Product Literature
Specifikationer
- Förstoring:100×/1,25 mm (i)
- Beskrivning:N-plan objective (oil/water)
- Seq:212