Orders LinkContactUsLinkComponent
VWR®, N-Plan Objectives
VWR®, N-Plan Objectives
N-plan objective, 20×/0,40 mm
Katalog # 630-2006
Leverantör:   VWR Collection
VWR®, N-Plan Objectives
Katalog # 630-2006
Leverantör:  VWR Collection
VWR®, N-Plan Objectives
VWR®, N-Plan Objectives
N-plan objective, 20×/0,40 mm
Price
760,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
These objectives are ideal for use in the education field and for laboratory routine applications.
IOS N-Plan objectives feature an infinity-corrected optical system.
Documents
Specifikationer
  • Förstoring:
    20×/0,40 mm
  • Beskrivning:
    N-plan objective
  • Seq:
    209