Micro protein quantification set, QuantiMate
Quantification set, 8,5 mm, Quartz
Catalog # 730-1453
Supplier: BIOCHROM
Jump to:
Product Details & Documents
QuantiMate is designed for concentration and purity determinations of nucleic acids and proteins.
- Measurement performance giving accurate and reproducible concentration results
- Samples measured without time consuming dilutions
- Only one cuvette is required
Dimensions are equivalent to standard cuvettes in order to work in most spectrophotometers.
Specifications
- Material:Quartz
- Beskrivning:Quantification set
- Höjd:8,5 mm
- Seq:001