Spray chambers and brackets
Bracket for mounting multi-mode sample introduction system (MSIS) on axial and radial ICP, for all 700 series
Katalog # VARI110868000
Leverantör: VARIAN
Gå till:
Produktdetaljer och dokument
These ICP-OES series are compatible with a range of accessories and supplies designed to extend the capabilities of the instruments.
Single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout.
Double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.
Sturman-Masters spray chamber is inert and minimizes interferences and improves stability with high TDS samples.
Specifikationer
- Beskrivning:Bracket for mounting multi-mode sample introduction system (MSIS) on axial and radial ICP, for all 700 series
- Seq:10