Sample dipper with v bottom
Dipper, wafer size 57 and smaller, square plate size 51 mm and smaller, max. thickness 1,5 mm
Katalog # ENTED11-0215
Leverantör: ENTEGRIS
Gå till:
Produktdetaljer och dokument
Holds a wafer or a square plate.
- Slot has 'V' bottom
- Clip on handle slides down to hold substrates in place
- Hold one wafer or one square plate
Dokument
ENTED15-0215_TDS_EN
Datablad
Specifikationer
- Beskrivning:Dipper, wafer size 57 and smaller, square plate size 51 mm and smaller, max. thickness 1,5 mm