Orders LinkContactUsLinkComponent
Pipette tips, ART™
Pipette tips, ART™
ART™ 10, 10 µl, +, 31,3 mm, Rack, 10 trays, 96 each
Catalog # 613-2461
   Low Manufacturing Impact
Pipette tips, ART™
Catalog # 613-2461
   Low Manufacturing Impact
Pipette tips, ART™
ART™ 10, 10 µl, +, 31,3 mm, Rack, 10 trays, 96 each
Price
608,00 kr
Sale
Valid until 31/03/2026
Log in for availability
1 * 960 Items
Product Details & Documents
These non filtered tips are designed to fit virtually any micropipette. They are suitable for use when working with the most difficult samples, such as radioactive compounds, viscous materials, fragile cell lines, viruses, difficult proteins and PCR reagents.
  • Non sterile tips are certified to be free of RNase, DNase, DNA, ATP, endotoxins, pyrogens and PCR inhibitors
  • Sterile tips are certified to be free of RNase, DNase, DNA, ATP, bioburden, endotoxins, pyrogens and PCR inhibitors
  • Rack cover can easily be realigned from a traditional lift-off to a hinged rack for one-handed access to tips
  • Front latch closes and locks ensure safe and secure storage
  • Space saving reload system is environmentally friendly, traceable and easy to use
Packaging: Tips are available in bulk packaging, in racks or in trays for rack reloading. Reload tips require an empty rack.
Specifications
  • Förp.:
    Rack, 10 trays, 96 each
  • Steril:
    +
  • Volym:
    10 µl
  • Längd:
    31,3 mm
  • Seq:
    05
  • Tip style:
    ART™ 10
  • Cat. No.:
    613-2461
  • Miljöcertificering:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Beskrivning
    Availability
    Price
  • 732-1170
    Tomma refillboxar, medium
    Log in for availability
    1 * 10 Items - 470,00 kr
Product Information
  • Catalog #
    Beskrivning
    Availability
    Price
  • 732-1169
    Tomma refillboxar, large
    Log in for availability
    1 * 8 Items - 195,00 kr
Product Information
  • Catalog #
    Beskrivning
    Availability
    Price
  • 732-1171
    Tomma refillboxar, small
    Log in for availability
    1 * 10 Items - 223,00 kr