Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Pipette tips, ART™

ART™ 200, 200 µl, -, 50,7 mm, Reload tower, 10 trays, 96 each
Katalog # 613-2496
   Low Manufacturing Impact
Leverantör:   Thermo Fisher Scientific
Pipette tips, ART™
Katalog # 613-2496
   Low Manufacturing Impact
Pipette tips, ART™

ART™ 200, 200 µl, -, 50,7 mm, Reload tower, 10 trays, 96 each

Price
573,00 kr
Kampanjpris

Gäller till kl 31/03/2026

Log in för att kolla tillgänglighet och ert pris
1 * 960 Items
Product Details & Documents

These non filtered tips are designed to fit virtually any micropipette. They are suitable for use when working with the most difficult samples, such as radioactive compounds, viscous materials, fragile cell lines, viruses, difficult proteins and PCR reagents.

  • Non sterile tips are certified to be free of RNase, DNase, DNA, ATP, endotoxins, pyrogens and PCR inhibitors
  • Sterile tips are certified to be free of RNase, DNase, DNA, ATP, bioburden, endotoxins, pyrogens and PCR inhibitors
  • Rack cover can easily be realigned from a traditional lift-off to a hinged rack for one-handed access to tips
  • Front latch closes and locks ensure safe and secure storage
  • Space saving reload system is environmentally friendly, traceable and easy to use

 : Tips are available in bulk packaging, in racks or in trays for rack reloading. Reload tips require an empty rack.

Specifikationer
  • Förp.:
    Reload tower, 10 trays, 96 each
  • Steril:
    -
  • Volym:
    200 µl
  • Längd:
    50,7 mm
  • Seq:
    16
  • Tip style:
    ART™ 200
  • Cat. No.:
    613-2496
  • Miljöcertificering:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories

Produktinformation

  • Katalog #
    Tillgänglighet
    Price
  • Tomma refillboxar, medium
    Tomma refillboxar, medium
    732-1170
    Log in för att kolla tillgänglighet och ert pris
    1 * 10 Items - 470,00 kr

Produktinformation

  • Katalog #
    Tillgänglighet
    Price
  • Tomma refillboxar, large
    Tomma refillboxar, large
    732-1169
    Log in för att kolla tillgänglighet och ert pris
    1 * 8 Items - 195,00 kr

Produktinformation

  • Katalog #
    Tillgänglighet
    Price
  • Tomma refillboxar, small
    Tomma refillboxar, small
    732-1171
    Log in för att kolla tillgänglighet och ert pris
    1 * 10 Items - 223,00 kr