Interfaces
Holder for Com.G@te® for Unistats® (bench top models)
Katalog # HUBE10019
Leverantör: HUBER
Gå till:
Product Details & Documents
Com.G@te®. The interfaces are integrated RS232 (bi-directional), RS485 (bi-directional), ECS external control signal, volt free contact, AIF analogue interface 0/4 to 20 mA or 0 to 10 V (bi-directional). Connections comply with NAMUR standard.
Specifikationer
- Beskrivning:Holder for Com.G@te® for Unistats® (bench top models)
- Seq:03