Brushes, FlexBrush™
Polyester cover, irradiated, NovaPoly
Katalog # 141-1644
Leverantör: MICRONOVA
Gå till:
Product Details & Documents
The FlexBrush™ is ideal for cleaning over and around piping and fittings. The flexibility of the head allows the user to shape the head for custom cleaning applications.
- Bendable brush with polyester or microfibre cover
- Acme threaded end attaches to a variety of handles for extended reach
- Threaded handle (STA series) can be added for extended reach
- Covers are compatible with strong disinfectants and autoclavable
Specifikationer
- Beskrivning:Polyester cover, irradiated, NovaPoly
- Seq:06
- Cat. No.:141-1644
- Environmentally Preferable:Waste Reducing