Orders LinkContactUsLinkComponent
Separating Funnels, Squibb
Separating Funnels, Squibb
500 ml, 29/32 NS
Katalog # KAVA632426355506
Leverantör:   Kavalier
Separating Funnels, Squibb
Katalog # KAVA632426355506
Leverantör:  Kavalier
Separating Funnels, Squibb
Separating Funnels, Squibb
500 ml, 29/32 NS
Price
3 225,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
Borosilicate 3.3 glass, delivered with NS-PE stopper.
  • Very good chemical and temperature resistence
  • Stopcock with PTFE key
  • Without graduations
Specifikationer
  • NS-hylsa:
    29/32 NS
  • Kapacitet:
    500 ml
  • Seq:
    04