Warning posts
General warning, red, PE
Catalog # 116-0635
Supplier: Brady
Jump to:
Product Details & Documents
These warning posts made of PE are ideal for controlling and directing foot traffic. Suitable for warehouses, factories, offices, etc. Chains and ribbons can be attached to the sturdy 90 cm high posts. The posts are available in one-colour or two-colour versions.
- Non conductive used for barricading electrically hazardous installations
- Built-in ultraviolet inhibitors block out ultraviolet rays to prevent colours from fading
- Colours are bright and eye-catching, even after years of use
Height: 900 mm
Specifications
- Beskrivning:General warning, red
- Material:PE
- Seq:02