Order Entry
Sweden
Orders LinkContactUsLinkComponent
Dialysis flasks, Slide-A-Lyzer™

Dialysis flasks, Slide-A-Lyzer™

Slide-A-Lyzer dialysis flasks, 3,5 kD, Rosa, 150 - 230 ml
Katalog # PIER87761
Leverantör:   Thermo Fisher Scientific
Dialysis flasks, Slide-A-Lyzer™
Katalog # PIER87761
Dialysis flasks, Slide-A-Lyzer™
Dialysis flasks, Slide-A-Lyzer™

Slide-A-Lyzer dialysis flasks, 3,5 kD, Rosa, 150 - 230 ml

Price
7 375,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 4 Items
Product Details & Documents

These dialysis flasks facilitate simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules in sample volumes up to 250 ml.

  • Easy to use
  • Color-coded for easy identification
  • No leakage, sample remains intact with no contamination
  • High surface area to volume ratio maximizes diffusion rate

Devices are constructed from two sheets of low protein-binding, regenerated cellulose membranes to ensure maximum sample recovery and purity and contain up to 85% less plastic per volume compared to other devices. Slide-A-Lyzer dialysis flasks are compatible with most laboratory buffers and can accomplish low-molecular weight contaminant removal, buffer exchange and desalting in 8 hours to overnight.

Specifikationer
  • MWCO:
    3,5 kD
  • Färg:
    Rosa
  • Beskrivning:
    Slide-A-Lyzer dialysis flasks
  • Seq:
    02
  • Provvolym:
    150 - 230 ml
Accessories

Produktinformation

  • Katalog #
    Tillgänglighet
    Price
  • Flotation disks for Slide-A-Lyzer™ dialysis flasks, LDPE
    Flotation disks for Slide-A-Lyzer™ dialysis flasks, LDPE
    PIER87759
    Log in för att kolla tillgänglighet och ert pris
    1 * 6 Items - 570,00 kr