Buffers, PIPES, 0,5 M solutions
PIPES buffer solution, 0,5 mol/L, 7,0, Glass bottle, 250 mL
Leverantör: Thermo Fisher Scientific
Gå till:
Product Details & Documents
Liquid
CAS No.: 5625-37-6
Appearance: Colorless
MDL No.: MFCD00006159
Caution: Storage Temperature: 2 to 8 °C
Documents
Specifikationer
Specifikationer
- Koncentration:0,5 mol/L
- Förpackningstyp:Glass bottle
- Förp.:250 mL
- Beskrivning:PIPES buffer solution
- pH-värde:7,0
- Cat. No.:J63234.AK
- CAS-nr:5625-37-6
- Förvaringstemperatur:2°C to 8°C
- Fysikalisk form:Liquid
- Utseende:Colourless
- MDL No.:MFCD00006159
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



