Buffers, PIPES, 0,5 M solutions
PIPES buffer solution, 0,5 mol/L, 8,0, Glass bottle, 100 mL
Leverantör: Thermo Fisher Scientific
Gå till:
Product Details & Documents
Liquid
CAS No.: 5625-37-6
Appearance: Colorless
MDL No.: MFCD00006159
Varning: Storage Temperature: 2 to 8 °C
Documents
Specifikationer
Specifikationer
- Koncentration:0,5 mol/L
- Förpackningstyp:Glass bottle
- Förp.:100 mL
- Beskrivning:PIPES buffer solution
- pH-värde:8,0
- Cat. No.:J61406.AE
- CAS-nr:5625-37-6
- Förvaringstemperatur:2°C to 8°C
- Fysikalisk form:Liquid
- Utseende:Colourless
- MDL No.:MFCD00006159



