Orders LinkContactUsLinkComponent
Loop interfaces, 4 – 20 mA current
Loop interfaces, 4 – 20 mA current
4 – 20 mA current loop interface, with temperature controller of 0 to +90 °C, UN30PA
Katalog # MEMMUN30PAV3
Leverantör:   MEMMERT
Loop interfaces, 4 – 20 mA current
Katalog # MEMMUN30PAV3
Leverantör:  MEMMERT
Loop interfaces, 4 – 20 mA current
Loop interfaces, 4 – 20 mA current
4 – 20 mA current loop interface, with temperature controller of 0 to +90 °C, UN30PA
Price
3 050,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivning:
    4 – 20 mA current loop interface, with temperature controller of 0 to +90 °C
  • Seq:
    072
  • För:
    UN30PA