VWR®, N-Plan Objectives
IOS N-planakromatiskt, 10×/0,25 mm
Katalog # 630-1640
Leverantör: VWR Collection
Gå till:
Produktdetaljer och dokument
These objectives are ideal for use in the education field and for laboratory routine applications.
IOS N-Plan objectives feature an infinity-corrected optical system.
Dokument
VWR® for Microscopy
Produktlitteratur
Specifikationer
- Förstoring:10×/0,25 mm
- Beskrivning:IOS N-planakromatiskt
- Seq:202