Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Ecamsule, Reference Standards

Ecamsule, USP reference standard, Vial, 25 mg
Katalog # 1231660.
Leverantör:   USP
Ecamsule, Reference Standards
Katalog # 1231660.
Leverantör:  USP
CAS-nummer:  
Ecamsule, Reference Standards

Ecamsule, USP reference standard, Vial, 25 mg

CAS-nummer:  
Price
13 400,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 25 mg
Produktdetaljer och dokument

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Specifikationer

Specifikationer

  • Förpackningstyp:
    Vial
  • Förp.:
    25 mg
  • Standard:
    USP reference standard
  • Beskrivning:
    Ecamsule
  • Cat. No.:
    1231660.
  • Benämning:
    Ecamsule
  • Synonym(s):
    4-((E)-((1SR,4SR)-7,7-dimethyl-3-oxo-4-(sulfomethyl)bicyclo[2.2.1]heptan-2-ylidene)methyl)benzoic acid

Specifikationer