Orders LinkContactUsLinkComponent
Separating funnels, Squibb
100 ml, 19/26 NS
Katalog # LENZ04018137
Leverantör:   Lenz Laborglas
Separating funnels, Squibb
Katalog # LENZ04018137
Leverantör:  Lenz Laborglas
Separating funnels, Squibb
Separating funnels, Squibb
100 ml, 19/26 NS
Price
3 550,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 2 Items
Product Details & Documents
Separating funnels, Squibb, stopcock with glass or PTFE key, made of borosilicate glass amberized.
  • Without graduation
  • Very good temperature and chemical resistance
  • Amber coloured for ligth sensitve liquids
Specifikationer
  • NS-hylsa:
    19/26 NS
  • Kapacitet:
    100 ml