D (+)-Mannos ≥98.0%
Glass bottle, 25 g
: TCI
: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
- :3458-28-4
- :C₆H₁₂O₆
- :MFCD00064122
- :180.16 g/mol
- :14,05747
- :376 °C (1013 hPa)
- :133 °C
- :1,539 g/cm³ (20 °C)
- :Omgivande
- :222-392-4
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:+14.0 to +15.5 deg (C=4, H2O)
- Solubility in Water:almost transparency
- Förpackningstyp:Glass bottle
- Förp.:25 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



