D (+)-Mannos ≥98.0%
Glass bottle, 500 g
Katalog # TCIAM0045-500G
Leverantör: TCI
Gå till:
Product Details & Documents
Synonymer: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
- CAS-nummer:3458-28-4
- Formel:C₆H₁₂O₆
- MDL-nummer:MFCD00064122
- Molecular Weight:180.16 g/mol
- Merck Index:14,05747
- Kokande Pt:376 °C (1013 hPa)
- Smält Pt:133 °C
- Densitet:1,539 g/cm³ (20 °C)
- Förvaringstemperatur:Omgivande
- EINECS:222-392-4
Documents
Specifikationer
Specifikationstestresultat
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Specific rotation [a]20/D:+14.0 to +15.5 deg (C=4, H2O)
- Solubility in Water:almost transparency
Specifikationer
- Förpackningstyp:Glass bottle
- Förp.:500 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



